Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Aminobenzoic Acid, 99%, ACROS Organics™

Click to view available options
Quantity:
250g
1kg
5kg
Packaging:
Glass bottle
Plastic drum
150-13-0
C7H7NO2
137.14
MFCD00007894
ALYNCZNDIQEVRV-UHFFFAOYSA-N
p-aminobenzoic acid,paba,para-aminobenzoic acid,vitamin bx,sunbrella,p-carboxyaniline,4-carboxyaniline,hachemina,paraminol,benzoic acid, 4-amino
978
CHEBI:30753
4-aminobenzoic acid
C1=CC(=CC=C1C(=O)O)N
150-13-0
C7H7NO2
137.14
MFCD00007894
ALYNCZNDIQEVRV-UHFFFAOYSA-N
p-aminobenzoic acid,paba,para-aminobenzoic acid,vitamin bx,sunbrella,p-carboxyaniline,4-carboxyaniline,hachemina,paraminol,benzoic acid, 4-amino
978
CHEBI:30753
4-aminobenzoic acid
C1=CC(=CC=C1C(=O)O)N
Color White to Yellow
Packaging Glass bottle
Quantity 250g
Melting Point 186.0°C to 189.0°C
Infrared Spectrum Authentic
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula H2NC6H4CO2H
Beilstein 14, 418
Fieser 02,24
Merck Index 15, 418
Solubility Information Solubility in water: 4.7g/L (20°C). Other solubilities: soluble in glacial acetic acid,soluble in ethyl acetate,slightly soluble in benzene,121g/L etanol- 1g/50mL ether,soluble in chloroform
Formula Weight 137.14
Physical Form Crystalline Powder
Percent Purity 99%
Chemical Name or Material 4-Aminobenzoic acid
Show More Show Less

GHS H Statement
Harmful to aquatic life with long lasting effects.

GHS P Statement
Avoid release to the environment.
Dispose of contents/container to an approved waste disposal plant.

Warning

EINECSNumber : 205-753-