Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Aminobenzoic acid (PABA), 98.5 to 101.5% (Dry Basis), Affymetrix/USB™

Click to view available options
Quantity:
200g
500g
1kg
5kg
Packaging:
Plastic bottle
150-13-0
C7H7NO2
137.138
MFCD00007894
ALYNCZNDIQEVRV-UHFFFAOYSA-N
p-aminobenzoic acid,paba,para-aminobenzoic acid,vitamin bx,sunbrella,p-carboxyaniline,4-carboxyaniline,hachemina,paraminol,benzoic acid, 4-amino
978
CHEBI:30753
4-aminobenzoic acid
C1=CC(=CC=C1C(=O)O)N
150-13-0
C7H7NO2
137.138
MFCD00007894
ALYNCZNDIQEVRV-UHFFFAOYSA-N
p-aminobenzoic acid,paba,para-aminobenzoic acid,vitamin bx,sunbrella,p-carboxyaniline,4-carboxyaniline,hachemina,paraminol,benzoic acid, 4-amino
978
CHEBI:30753
4-aminobenzoic acid
C1=CC(=CC=C1C(=O)O)N
Formula Weight 137.14
Physical Form Crystalline Powder
Percent Purity 98.5 to 101.5%
Packaging Plastic bottle
Specific Gravity 1.374
Melting Point ~187°C (decomposition)
Quantity 500g
Chemical Name or Material 4-Aminobenzoic acid (PABA)
Assay Dry basis