Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Aminobenzyl alcohol, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
50g
Packaging:
Glass bottle
623-04-1
C7H9NO
123.15
MFCD00014782
AXKGIPZJYUNAIW-UHFFFAOYSA-N
4-aminobenzyl alcohol,4-aminophenyl methanol,4-aminobenzylalcohol,4-hydroxymethyl aniline,p-aminobenzyl alcohol,p-aminobenzylalcohol,benzenemethanol, 4-amino,benzyl alcohol, p-amino,4-amino-benzylalcohol,pubchem7480
69331
(4-aminophenyl)methanol
C1=CC(=CC=C1CO)N
623-04-1
C7H9NO
123.15
MFCD00014782
AXKGIPZJYUNAIW-UHFFFAOYSA-N
4-aminobenzyl alcohol,4-aminophenyl methanol,4-aminobenzylalcohol,4-hydroxymethyl aniline,p-aminobenzyl alcohol,p-aminobenzylalcohol,benzenemethanol, 4-amino,benzyl alcohol, p-amino,4-amino-benzylalcohol,pubchem7480
69331
(4-aminophenyl)methanol
C1=CC(=CC=C1CO)N
Melting Point 60.0°C to 63.0°C
Quantity 5g
Infrared Spectrum Authentic
Color Yellow to Brown
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (HPLC)
Formula Weight 123.15
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material 4-Aminobenzyl alcohol, 99%
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER

GHS Signal Word: Warning

EINECSNumber : 210-767-5