Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Aminobutyraldehyde Dimethyl Acetal 98.0+%, TCI America™

Click to view available options
Quantity:
25mL
500mL
19060-15-2
C6H15NO2
133.191
MFCD00189373
TYVAXMOICMBSMT-UHFFFAOYSA-N
4-aminobutyraldehyde dimethyl acetal,4,4-dimethoxybutylamine,4,4-dimethoxy-1-butanamine,4-aminobutyraldehyde dimethylacetal,acmc-1bqj6,4-aminobutyraldehyde dimethyl,1-amino-4,4-dimethoxybutane,cs-waa0041,tyvaxmoicmbsmt-uhfffaoysa,4-aminobutyraldehydedimethylacetal
5463803
4,4-dimethoxybutan-1-amine
COC(CCCN)OC
19060-15-2
C6H15NO2
133.191
MFCD00189373
TYVAXMOICMBSMT-UHFFFAOYSA-N
4-aminobutyraldehyde dimethyl acetal,4,4-dimethoxybutylamine,4,4-dimethoxy-1-butanamine,4-aminobutyraldehyde dimethylacetal,acmc-1bqj6,4-aminobutyraldehyde dimethyl,1-amino-4,4-dimethoxybutane,cs-waa0041,tyvaxmoicmbsmt-uhfffaoysa,4-aminobutyraldehydedimethylacetal
5463803
4,4-dimethoxybutan-1-amine
COC(CCCN)OC
Boiling Point 85°C
Quantity 500mL
Color Yellow
UN Number 2924
Formula Weight 133.19
Physical Form Clear Liquid at 20°C
Percent Purity ≥98.0% (GC,T)
Chemical Name or Material 4-Aminobutyraldehyde Dimethyl Acetal