Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Aminobutyric Acid, Reagent, 98%, Spectrum™

Click to view available options
Quantity:
25 g
100 g
500 g
12 kg
25 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
56-12-2
C4H9NO2
103.12
BTCSSZJGUNDROE-UHFFFAOYSA-N
4-aminobutanoic acid
NCCCC(O)=O
56-12-2
C4H9NO2
103.12
BTCSSZJGUNDROE-UHFFFAOYSA-N
4-aminobutanoic acid
NCCCC(O)=O

AM151, 56-12-2, C4H9NO2

Spectrum™ 4-Aminobutyric Acid, Reagent belongs to the transaminases family which transfer nitrogenous groups. It is an enzyme which plays a part in 5 metabolic pathways: glutamate metabolism, beta-alanine metabolism, alanine and aspartate metabolism, butanoate metabolism, and propionate metabolism. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.
Ignition Residue 0.33%
Loss on Drying 0.5%
Packaging Poly Bottle
Quantity 100 g
CAS Max % 100%
Percent Purity 98%