Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Aminocarbonyl-3-nitrobenzoic acid, 98%

Click to view available options
Quantity:
1g
5g
25g
87594-59-0
C8H6N2O5
210.145
MFCD04039778
LKPGLAHLFCDSNK-UHFFFAOYSA-N
4-carbamoyl-3-nitrobenzoic acid,2-nitro-4-carboxybenzamide,4-carbamoyl-3-nitro-benzoic acid,pubchem13255,3-nitroterephthalamic acid,4-carboxy-2-nitrobenzamide,4-aminocarbonyl-3-nitrobenzoic acid,4-aminocarbonyl-3-nitro-benzoic acid
7016425
C1=CC(=C(C=C1C(=O)O)[N+](=O)[O-])C(=O)N
87594-59-0
C8H6N2O5
210.145
MFCD04039778
LKPGLAHLFCDSNK-UHFFFAOYSA-N
4-carbamoyl-3-nitrobenzoic acid,2-nitro-4-carboxybenzamide,4-carbamoyl-3-nitro-benzoic acid,pubchem13255,3-nitroterephthalamic acid,4-carboxy-2-nitrobenzamide,4-aminocarbonyl-3-nitrobenzoic acid,4-aminocarbonyl-3-nitro-benzoic acid
7016425
C1=CC(=C(C=C1C(=O)O)[N+](=O)[O-])C(=O)N
Quantity 1g
Formula Weight 210.15
Percent Purity 98%
Chemical Name or Material 4-Aminocarbonyl-3-nitrobenzoic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning