Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Aminoimidazole-5-carboxamide, 95%

Click to view available options
Quantity:
5g
25g
100g
360-97-4
C4H6N4O
126.119
MFCD02181040
DVNYTAVYBRSTGK-UHFFFAOYSA-N
5-aminoimidazole-4-carboxamide,5-amino-1h-imidazole-4-carboxamide,4-amino-5-imidazolecarboxamide,5-amino-4-imidazolecarboxamide,colahepat,4-aminoimidazole-5-carboxamide,aica,1h-imidazole-4-carboxamide, 5-amino,diazol-c,4-carbamoyl-5-aminoimidazole
9679
CHEBI:2030
4-amino-1H-imidazole-5-carboxamide
C1=NC(=C(N1)C(=O)N)N
360-97-4
C4H6N4O
126.119
MFCD02181040
DVNYTAVYBRSTGK-UHFFFAOYSA-N
5-aminoimidazole-4-carboxamide,5-amino-1h-imidazole-4-carboxamide,4-amino-5-imidazolecarboxamide,5-amino-4-imidazolecarboxamide,colahepat,4-aminoimidazole-5-carboxamide,aica,1h-imidazole-4-carboxamide, 5-amino,diazol-c,4-carbamoyl-5-aminoimidazole
9679
CHEBI:2030
4-amino-1H-imidazole-5-carboxamide
C1=NC(=C(N1)C(=O)N)N
Applications
4-Aminoimidazole-5-carboxamide is used as Catalytic agent, Petrochemical additive. It is metabolite of temozolomide.

Solubility
Soluble in DMSO, Methanol, Water.

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
Quantity 100g
Melting Point 164°C to 170°C
Color White
Beilstein 3938
Formula Weight 126.12
Physical Form Powder
Percent Purity 95%
Chemical Name or Material 4-Aminoimidazole-5-carboxamide

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : NI3910000

TSCA : TSCA