Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Aminoimidazole-5-carboxamide hydrochloride, 98%

Click to view available options
Quantity:
1g
5g
25g
72-40-2
C4H7ClN4O
162.577
MFCD00012704
MXCUYSMIELHIQL-UHFFFAOYSA-N
4-amino-5-imidazolecarboxamide hydrochloride,5-amino-4-imidazolecarboxamide hydrochloride,5-amino-1h-imidazole-4-carboxamide hydrochloride,5-aminoimidazole-4-carboxamide hydrochloride,aic .cntdot. hcl,4-amino-1h-imidazole-5-carboxamide hydrochloride,aminoimidazolecarboxamide hydrochloride,4-aminoimidazole-5-carboxamide hydrochoride,1h-imidazole-4-carboxamide, 5-amino-, monohydrochloride,5-aminoimidazol-4-carboxamide, hydrochloride
66146
4-amino-1H-imidazole-5-carboxamide;hydrochloride
C1=NC(=C(N1)C(=O)N)N.Cl
72-40-2
C4H7ClN4O
162.577
MFCD00012704
MXCUYSMIELHIQL-UHFFFAOYSA-N
4-amino-5-imidazolecarboxamide hydrochloride,5-amino-4-imidazolecarboxamide hydrochloride,5-amino-1h-imidazole-4-carboxamide hydrochloride,5-aminoimidazole-4-carboxamide hydrochloride,aic .cntdot. hcl,4-amino-1h-imidazole-5-carboxamide hydrochloride,aminoimidazolecarboxamide hydrochloride,4-aminoimidazole-5-carboxamide hydrochoride,1h-imidazole-4-carboxamide, 5-amino-, monohydrochloride,5-aminoimidazol-4-carboxamide, hydrochloride
66146
4-amino-1H-imidazole-5-carboxamide;hydrochloride
C1=NC(=C(N1)C(=O)N)N.Cl
Quantity 1g
Odor Odorless
Melting Point ~270°C (decomposition)
Beilstein 3701645
Formula Weight 162.58
Percent Purity 98%
Chemical Name or Material 4-Aminoimidazole-5-carboxamide hydrochloride

RTECSNumber : NI3911000

TSCA : TSCA