Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Aminomethyl-2-chloropyridine, 95%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
144900-57-2
C6H7ClN2
142.59
GGHCWJWUOSNCSK-UHFFFAOYSA-N
2-chloropyridin-4-yl methanamine,4-pyridinemethanamine, 2-chloro,2-chloropyridin-4-yl methylamine,4-aminomethyl-2-chloropyridine,2-chloro-4-aminomethylpyridine,2-chloro-4-pyridyl methylamine,1-2-chloro-4-pyridinyl methanamine,2-chloro-4-pyridinemethanamine,1-2-chloropyridin-4-yl methanamine,c-2-chloro-pyridin-4-yl-methylamine
2794167
(2-chloropyridin-4-yl)methanamine
C1=CN=C(C=C1CN)Cl
144900-57-2
C6H7ClN2
142.59
GGHCWJWUOSNCSK-UHFFFAOYSA-N
2-chloropyridin-4-yl methanamine,4-pyridinemethanamine, 2-chloro,2-chloropyridin-4-yl methylamine,4-aminomethyl-2-chloropyridine,2-chloro-4-aminomethylpyridine,2-chloro-4-pyridyl methylamine,1-2-chloro-4-pyridinyl methanamine,2-chloro-4-pyridinemethanamine,1-2-chloropyridin-4-yl methanamine,c-2-chloro-pyridin-4-yl-methylamine
2794167
(2-chloropyridin-4-yl)methanamine
C1=CN=C(C=C1CN)Cl
Color Colorless to Yellow
Quantity 5g
Infrared Spectrum Authentic
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 94% min. (GC)
Formula Weight 142.59
Physical Form Liquid
Percent Purity 95%
Chemical Name or Material 4-Aminomethyl-2-chloropyridine

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes severe skin burns and eye damage.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger