Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-(Aminomethyl)benzoic acid, 97%

Click to view available options
Quantity:
25g
100g
500g
56-91-7
C8H9NO2
151.165
MFCD00010203
QCTBMLYLENLHLA-UHFFFAOYSA-N
4-aminomethyl benzoic acid,4-carboxybenzylamine,pamba,styptopur,gumbix,p-aminomethylbenzoic acid,aminomethylbenzoic acid,benzoic acid, 4-aminomethyl,benzylamine-4-carboxylic acid,alpha-amino-p-toluic acid
65526
4-(aminomethyl)benzoic acid
C1=CC(=CC=C1CN)C(=O)O
56-91-7
C8H9NO2
151.165
MFCD00010203
QCTBMLYLENLHLA-UHFFFAOYSA-N
4-aminomethyl benzoic acid,4-carboxybenzylamine,pamba,styptopur,gumbix,p-aminomethylbenzoic acid,aminomethylbenzoic acid,benzoic acid, 4-aminomethyl,benzylamine-4-carboxylic acid,alpha-amino-p-toluic acid
65526
4-(aminomethyl)benzoic acid
C1=CC(=CC=C1CN)C(=O)O
Applications
4-(Aminomethyl)benzoic acid acts as an unnatural amino acid derivative. It is also used as a type 2 antifibrinolytic agent. Further, it reacts with 2-methyl-isothiourea sulfate to prepare 4-guanidinomethylbenzoic acid.

Solubility
Slightly soluble in water. Insoluble in ethanol, benzene and chloroform.

Notes
Incompatible with strong oxidizing agents, strong reducing agents and strong bases.
Quantity 25g
Melting Point ~290°C (decomposition)
Beilstein 1100606
Formula Weight 151.16
Percent Purity 97%
Chemical Name or Material 4-(Aminomethyl)benzoic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning