Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
4-(Aminomethyl)pyridine, 98%, Acros Organics
Click to view available options
Quantity:
25g
100g
500g
Specifications
Specific Gravity | 1.065 |
Quantity | 25g |
Packaging | Glass bottle |
Density | 1.0650g/mL |
Melting Point | -8.0°C |
Boiling Point | 228.0°C to 230.0°C |
Flash Point | 108°C |
Refractive Index | 1.5515 |
Assay Percent Range | 97.5% min. (GC) |
Formula Weight | 108.14 |
Show More |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately
EINECSNumber : 223-092-6