Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Aminomethyl)pyridine, 98%, Acros Organics

Click to view available options
Quantity:
25g
100g
500g
3731-53-1
C6H8N2
108.14
TXQWFIVRZNOPCK-UHFFFAOYSA-N
4-pyridinemethaneamine,4-aminomethyl pyridine,4-pyridinemethanamine,4-pyridylmethylamine,4-picolylamine,4-aminomethylpyridine,4-picolinylamine,4-pyridinemethylamine,1-pyridin-4-ylmethanamine,4-picolinamine
77317
pyridin-4-ylmethanamine
C1=CN=CC=C1CN
3731-53-1
C6H8N2
108.14
TXQWFIVRZNOPCK-UHFFFAOYSA-N
4-pyridinemethaneamine,4-aminomethyl pyridine,4-pyridinemethanamine,4-pyridylmethylamine,4-picolylamine,4-aminomethylpyridine,4-picolinylamine,4-pyridinemethylamine,1-pyridin-4-ylmethanamine,4-picolinamine
77317
pyridin-4-ylmethanamine
C1=CN=CC=C1CN
Flash Point 108°C
Specific Gravity 1.065
Boiling Point 228.0°C to 230.0°C
Quantity 500g
Packaging Glass bottle
Density 1.0650g/mL
Melting Point -8.0°C
Refractive Index 1.5515
Assay Percent Range 97.5% min. (GC)
Formula Weight 108.14
Percent Purity 98%
Chemical Name or Material 4-(Aminomethyl)pyridine
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately

EINECSNumber : 223-092-6