Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
4-Aminomethyltetrahydropyran, 97%, Acros Organics
Click to view available options
Quantity:
250mg
1g
5g
25g
Specifications
Specific Gravity | 1.02 |
Density | 1.0200g/mL |
Packaging | Glass bottle |
Refractive Index | 1.4640 to 1.4660 |
Boiling Point | 75.0°C to 76.0°C (18.0mmHg) |
Quantity | 5g |
Assay Percent Range | 96% min. (GC) |
Formula Weight | 115.17 |
Percent Purity | 97% |
Chemical Name or Material | 4-Aminomethyltetrahydropyran |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger