Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Aminonicotinic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
7418-65-7
C6H6N2O2
138.13
MFCD00234183
IASBMUIXBJNMDW-UHFFFAOYSA-N
4-aminonicotinic acid,4-amino-3-pyridinecarboxylic acid,4-amino-nicotinic acid,4-amino-3-carboxypyridine,4-aminonicotinicacid,4-amino nicotinic acid,3-pyridinecarboxylic acid, 4-amino,4-amino-3-pyridine carboxylic acid,zlchem 370,pubchem1214
319979
CHEBI:68577
4-aminopyridine-3-carboxylic acid
C1=CN=CC(=C1N)C(=O)O
7418-65-7
C6H6N2O2
138.13
MFCD00234183
IASBMUIXBJNMDW-UHFFFAOYSA-N
4-aminonicotinic acid,4-amino-3-pyridinecarboxylic acid,4-amino-nicotinic acid,4-amino-3-carboxypyridine,4-aminonicotinicacid,4-amino nicotinic acid,3-pyridinecarboxylic acid, 4-amino,4-amino-3-pyridine carboxylic acid,zlchem 370,pubchem1214
319979
CHEBI:68577
4-aminopyridine-3-carboxylic acid
C1=CN=CC(=C1N)C(=O)O
Infrared Spectrum Authentic
Color Cream to Yellow
Quantity 5g
Packaging Glass bottle
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 138.13
Physical Form Powder
Percent Purity 97%
Chemical Name or Material 4-Aminonicotinic acid
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.