Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Aminophthalic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
5434-21-9
C8H7NO4
181.15
MFCD00013985
OXSANYRLJHSQEP-UHFFFAOYSA-N
4-aminobenzene-1,2-dicarboxylic acid,1,2-benzenedicarboxylic acid, 4-amino,4-amino-1,2-benzenedicarboxylic acid,4-amino phthalic acid,contains 3-aminophthalic acid,pubchem16853,acmc-1arqu,4-aminophthalic acid,ksc270i4n,ghl.pd_mitscher_leg0.1208
72912
4-aminophthalic acid
C1=CC(=C(C=C1N)C(=O)O)C(=O)O
5434-21-9
C8H7NO4
181.15
MFCD00013985
OXSANYRLJHSQEP-UHFFFAOYSA-N
4-aminobenzene-1,2-dicarboxylic acid,1,2-benzenedicarboxylic acid, 4-amino,4-amino-1,2-benzenedicarboxylic acid,4-amino phthalic acid,contains 3-aminophthalic acid,pubchem16853,acmc-1arqu,4-aminophthalic acid,ksc270i4n,ghl.pd_mitscher_leg0.1208
72912
4-aminophthalic acid
C1=CC(=C(C=C1N)C(=O)O)C(=O)O
Melting Point 344°C
Quantity 1g
Packaging Glass bottle
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula NH2C6H3(COOH)2
Formula Weight 181.15
Percent Purity 97%
Chemical Name or Material 4-Aminophthalic acid

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 226-596-4