Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Aminopyridine-2-carboxylic acid, 97%, Acros Organics™

Click to view available options
Quantity:
250mg
1g
100047-36-7
C6H6N2O2
138.12
JRZBTJVSAANBEV-UHFFFAOYSA-N
4-aminopicolinic acid,4-aminopyridine-2-carboxylicacid,2-pyridinecarboxylic acid, 4-amino,4-amino-2-pyridinecarboxylic acid,4-amino-pyridine-2-carboxylic acid,4-aminopicolinicacid,4-amino-picolinsaure,pubchem22241,2-carboxy-4-aminopyridine,picolinic acid, 4-amino
2050132
4-aminopyridine-2-carboxylic acid
C1=CN=C(C=C1N)C(=O)O
100047-36-7
C6H6N2O2
138.12
JRZBTJVSAANBEV-UHFFFAOYSA-N
4-aminopicolinic acid,4-aminopyridine-2-carboxylicacid,2-pyridinecarboxylic acid, 4-amino,4-amino-2-pyridinecarboxylic acid,4-amino-pyridine-2-carboxylic acid,4-aminopicolinicacid,4-amino-picolinsaure,pubchem22241,2-carboxy-4-aminopyridine,picolinic acid, 4-amino
2050132
4-aminopyridine-2-carboxylic acid
C1=CN=C(C=C1N)C(=O)O
Packaging Glass Bottle
Melting Point 319.0°C
Quantity 1g
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Formula Weight 138.12
Percent Purity 97%
Chemical Name or Material 4-Aminopyridine-2-carboxylic acid

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

GHS Signal Word: Warning