Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Aminopyrimidine, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
591-54-8
C4H5N3
95.1
MFCD00006112
OYRRZWATULMEPF-UHFFFAOYSA-N
4-aminopyrimidine,4-pyrimidinamine,4-pyrimidineamine,pyrimidin-4-ylamine,pyrimidine, 4-amino,6-aminopyrimidine,4-amino pyrimidine,pyrimidine-4-ylamine,pyrimidin-4-yl-amine,4-pyrimidinamine 9ci
68958
CHEBI:38616
pyrimidin-4-amine
C1=CN=CN=C1N
591-54-8
C4H5N3
95.1
MFCD00006112
OYRRZWATULMEPF-UHFFFAOYSA-N
4-aminopyrimidine,4-pyrimidinamine,4-pyrimidineamine,pyrimidin-4-ylamine,pyrimidine, 4-amino,6-aminopyrimidine,4-amino pyrimidine,pyrimidine-4-ylamine,pyrimidin-4-yl-amine,4-pyrimidinamine 9ci
68958
CHEBI:38616
pyrimidin-4-amine
C1=CN=CN=C1N
Quantity 1g
Melting Point 151°C to 155°C
Color White to Beige
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (HPLC)
Formula Weight 95.1
Physical Form Crystals or Powder
Percent Purity 98%
Chemical Name or Material 4-Aminopyrimidine
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning