Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Aminoquinazoline, 97%

Click to view available options
Quantity:
1g
5g
15018-66-3
C8H7N3
145.165
MFCD00092016
DRYRBWIFRVMRPV-UHFFFAOYSA-N
quinazolin-4-ylamine,4-quinazolinamine,4-aminoquinazoline,4-quinazolineamine,quinazoline, 4-amino,quinazoline-4-ylamine,1lq,4-amino quinazoline,4-amino-quinazoline,4-quinazolinamine #
84759
quinazolin-4-amine
C1=CC=C2C(=C1)C(=NC=N2)N
15018-66-3
C8H7N3
145.165
MFCD00092016
DRYRBWIFRVMRPV-UHFFFAOYSA-N
quinazolin-4-ylamine,4-quinazolinamine,4-aminoquinazoline,4-quinazolineamine,quinazoline, 4-amino,quinazoline-4-ylamine,1lq,4-amino quinazoline,4-amino-quinazoline,4-quinazolinamine #
84759
quinazolin-4-amine
C1=CC=C2C(=C1)C(=NC=N2)N
Quantity 5g
Beilstein 118621
Formula Weight 145.06
Physical Form Solid
Percent Purity 97%
Chemical Name or Material 4-Aminoquinazoline

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : VA1236000