Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Aminosalicylic acid, 98+%

Click to view available options
Quantity:
100g
500g
65-49-6
C7H7NO3
153.137
MFCD00007789
WUBBRNOQWQTFEX-UHFFFAOYSA-N
4-aminosalicylic acid,aminosalicylic acid,p-aminosalicylic acid,rezipas,pamisyl,parasalindon,aminopar,deapasil,ferrosan,apacil
4649
CHEBI:27565
4-amino-2-hydroxybenzoic acid
C1=CC(=C(C=C1N)O)C(=O)O
65-49-6
C7H7NO3
153.137
MFCD00007789
WUBBRNOQWQTFEX-UHFFFAOYSA-N
4-aminosalicylic acid,aminosalicylic acid,p-aminosalicylic acid,rezipas,pamisyl,parasalindon,aminopar,deapasil,ferrosan,apacil
4649
CHEBI:27565
4-amino-2-hydroxybenzoic acid
C1=CC(=C(C=C1N)O)C(=O)O
Applications
Competitive pteridine synthetase inhibitor, has antibacterial capable of inhibiting the growth of Mycobacterium tuberculosis. It inhibits NF-κB and folic acid synthesis.

Solubility
Soluble in water (2 g/L) at 20°C, dilute sodium hydroxide, dilute nitric acid, ether (slightly), methanol, acetone, and ethanol. Virtually insoluble in benzene, chloroform, and carbon tetrachloride.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
Melting Point ~135°C to 145°C (decomposition)
Odor Odorless
Quantity 500g
Beilstein 473071
Merck Index 14,477
Formula Weight 153.14
Percent Purity ≥98%
Chemical Name or Material 4-Aminosalicylic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : VO1225000

TSCA : TSCA