Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Aminosalicylic Acid Calcium Salt Heptahydrate 98.0+%, TCI America™

Click to view available options
Quantity:
25g
500g
133-15-3
C14H12CaN2O6
344.336
MFCD00067430
XDWVNCOPMIEDJK-UHFFFAOYSA-L
4-Amino-2-hydroxybenzoic Acid Calcium Salt, Dicalcium Bis(4-aminosalicylate)
102600765
calcium;4-amino-2-hydroxybenzoate
C1=CC(=C(C=C1N)O)C(=O)[O-].C1=CC(=C(C=C1N)O)C(=O)[O-].[Ca+2]
133-15-3
C14H12CaN2O6
344.336
MFCD00067430
XDWVNCOPMIEDJK-UHFFFAOYSA-L
4-Amino-2-hydroxybenzoic Acid Calcium Salt, Dicalcium Bis(4-aminosalicylate)
102600765
calcium;4-amino-2-hydroxybenzoate
C1=CC(=C(C=C1N)O)C(=O)[O-].C1=CC(=C(C=C1N)O)C(=O)[O-].[Ca+2]
Color Undesignated
Quantity 25g
Formula Weight 382.40
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (T)
Chemical Name or Material 4-Aminosalicylic Acid Calcium Salt Heptahydrate

RTECSNumber : VO1470000

TSCA : No