Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amylbenzoic Acid 98.0+%, TCI America™

Click to view available options
Quantity:
5g
25g
250g
26311-45-5
C12H16O2
192.258
MFCD00002572
CWYNKKGQJYAHQG-UHFFFAOYSA-N
benzoic acid, 4-pentyl,4-n-pentylbenzoic acid,p-pentylbenzoic acid,4-amylbenzoic acid,p-amylbenzoic acid,para-pentylbenzoic acid,benzoic acid, p-pentyl,4-pentyl benzoic acid,p-n-pentylbenzoic acid,4-pentylbenzoicacid
33479
4-pentylbenzoic acid
CCCCCC1=CC=C(C=C1)C(=O)O
26311-45-5
C12H16O2
192.258
MFCD00002572
CWYNKKGQJYAHQG-UHFFFAOYSA-N
benzoic acid, 4-pentyl,4-n-pentylbenzoic acid,p-pentylbenzoic acid,4-amylbenzoic acid,p-amylbenzoic acid,para-pentylbenzoic acid,benzoic acid, p-pentyl,4-pentyl benzoic acid,p-n-pentylbenzoic acid,4-pentylbenzoicacid
33479
4-pentylbenzoic acid
CCCCCC1=CC=C(C=C1)C(=O)O
Quantity 25g
Color White
Melting Point 89°C
Formula Weight 192.26
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (T)
Chemical Name or Material 4-Amylbenzoic Acid

EINECSNumber : (3)-1338

RTECSNumber : DH6273000

TSCA : Yes