Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amyloxycinnamic acid, 97%, Acros Organics

Click to view available options
Quantity:
1g
62718-63-2
C14H18O3
234.295
MFCD00037797
WXPBGFBUVHLMSK-JXMROGBWSA-N
p-pentyloxycinnamic acid,3-4-pentyloxy phenyl acrylic acid,4-amyloxycinnamic acid,3-4-pentyloxyphenyl-2-propenoic acid,2e-3-4-pentyloxy phenyl prop-2-enoic acid,2e-3-4-pentyloxyphenyl prop-2-enoic acid,4-pentoxycinnamic acid,e-3-4-pentyloxy phenyl acrylic acid,3-4-pentyloxy phenyl acrylicacid,e-3-4-pentoxyphenyl prop-2-enoic acid
1715219
(E)-3-(4-pentoxyphenyl)prop-2-enoic acid
CCCCCOC1=CC=C(C=C1)C=CC(=O)O
62718-63-2
C14H18O3
234.295
MFCD00037797
WXPBGFBUVHLMSK-JXMROGBWSA-N
p-pentyloxycinnamic acid,3-4-pentyloxy phenyl acrylic acid,4-amyloxycinnamic acid,3-4-pentyloxyphenyl-2-propenoic acid,2e-3-4-pentyloxy phenyl prop-2-enoic acid,2e-3-4-pentyloxyphenyl prop-2-enoic acid,4-pentoxycinnamic acid,e-3-4-pentyloxy phenyl acrylic acid,3-4-pentyloxy phenyl acrylicacid,e-3-4-pentoxyphenyl prop-2-enoic acid
1715219
(E)-3-(4-pentoxyphenyl)prop-2-enoic acid
CCCCCOC1=CC=C(C=C1)C=CC(=O)O
Quantity 1g
Color White
Melting Point 138°C to 145°C
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula CH3(CH)4OC6H4(CH)COOH
Beilstein 10, IV, 1005
Formula Weight 234.29
Physical Form Crystals or Needle-Like Powder
Percent Purity ≥96.0%
Chemical Name or Material 4-Amyloxycinnamic acid, 97%
Show More Show Less