Alfa Aesar™ 4-Anilino-1-benzylpiperidine, 98%

Manufacturer: Alfa Aesar™ H2715814

Quantity 25g

 View more versions of this product

Catalog No. AAH2715814

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1155-56-2
    Molecular Formula C18H22N2
    Formula Weight 266.38
    MDL Number MFCD00023738
    Synonym 1-Benzyl-N-phenyl-4-piperidinamine
    Chemical Name or Material 4-Anilino-1-benzylpiperidine

    Specifications

    Formula Weight 266.38
    Molecular Formula C18H22N2
    MDL Number MFCD00023738
    CAS 1155-56-2
    Synonym 1-Benzyl-N-phenyl-4-piperidinamine
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Quantity 25g
    Health Hazard 3 GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Chemical Name or Material 4-Anilino-1-benzylpiperidine
    Assay 98%

    Description

    Applications
    4-Anilino-1-benzylpiperidine, is used as a pharmaceutical intermediate.

    Solubility
    Soluble in dichloromethane.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.

    GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit