Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Azabenzimidazole, 99%

Click to view available options
Quantity:
1g
5g
25g
273-21-2
C6H5N3
119.127
MFCD00005579
GAMYYCRTACQSBR-UHFFFAOYSA-N
3h-imidazo 4,5-b pyridine,4-azabenzimidazole,1h-imidazo 4,5-b pyridine,1-deazapurine,3,4-diazaindole,7-azabenzimidazole,imidazopyridine,4h-imidazo 4,5-b pyridine,pyrido 2,3-d imidazole,azabenzimidazole
67504
CHEBI:59952
1H-imidazo[4,5-b]pyridine
C1=CC2=C(N=C1)N=CN2
273-21-2
C6H5N3
119.127
MFCD00005579
GAMYYCRTACQSBR-UHFFFAOYSA-N
3h-imidazo 4,5-b pyridine,4-azabenzimidazole,1h-imidazo 4,5-b pyridine,1-deazapurine,3,4-diazaindole,7-azabenzimidazole,imidazopyridine,4h-imidazo 4,5-b pyridine,pyrido 2,3-d imidazole,azabenzimidazole
67504
CHEBI:59952
1H-imidazo[4,5-b]pyridine
C1=CC2=C(N=C1)N=CN2
Applications
4-Azabenzimidazole is used in the preparation of imidazo[4,5-b]pyridine 4-oxide. It is also used as a reagent to prepare benzoylmethyl-substituted azoles which are inhibitors of nitric oxide synthase with antioxidant properties.

Solubility
Soluble in dimethylformamide.

Notes
Incompatible with strong oxidizing agent and strong acid.
Boiling Point 140°C to 150°C (0.1mmHg)
Melting Point 145°C to 150°C
Quantity 25g
Beilstein 2600
Formula Weight 119.13
Percent Purity 99%
Chemical Name or Material 4-Azabenzimidazole

RTECSNumber : NJ5108000