Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Azepanone hydrochloride, 96%

Click to view available options
Quantity:
1g
5g
25g
50492-22-3
C6H12ClNO
149.618
MFCD02183487
WFTRLIZPJMFJER-UHFFFAOYSA-N
4-perhydroazepinone hydrochloride,azepan-4-one hydrochloride,hexahydro-4h-azepin-4-one hydrochloride,4-azepanone hydrochloride,hexahydro-4-azepinone hydrochloride,4-perhydroazepinone hcl,4h-azepin-4-one, hexahydro-, hydrochloride,hexahydro-4h-azepin-4-one hcl,4-oxohomopiperidine-hcl,pubchem17662
22021728
azepan-4-one;hydrochloride
C1CC(=O)CCNC1.Cl
50492-22-3
C6H12ClNO
149.618
MFCD02183487
WFTRLIZPJMFJER-UHFFFAOYSA-N
4-perhydroazepinone hydrochloride,azepan-4-one hydrochloride,hexahydro-4h-azepin-4-one hydrochloride,4-azepanone hydrochloride,hexahydro-4-azepinone hydrochloride,4-perhydroazepinone hcl,4h-azepin-4-one, hexahydro-, hydrochloride,hexahydro-4h-azepin-4-one hcl,4-oxohomopiperidine-hcl,pubchem17662
22021728
azepan-4-one;hydrochloride
C1CC(=O)CCNC1.Cl
Melting Point 176°C to 181°C
Quantity 25g
Beilstein 3687441
Formula Weight 149.62
Physical Form Solid
Percent Purity 96%
Chemical Name or Material 4-Azepanone hydrochloride