Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Azidocinnamaldehyde 98.0+%, TCI America™

Click to view available options
Quantity:
5g
22736-78-3
C9H7N3O
173.175
MFGPIUGUWLAIGM-UHFFFAOYSA-N
3-(4-Azidophenyl)acrylaldehyde, 3-(4-Azidophenyl)-2-propenal
44629867
3-(4-azidophenyl)prop-2-enal
C1=CC(=CC=C1C=CC=O)N=[N+]=[N-]
22736-78-3
C9H7N3O
173.175
MFGPIUGUWLAIGM-UHFFFAOYSA-N
3-(4-Azidophenyl)acrylaldehyde, 3-(4-Azidophenyl)-2-propenal
44629867
3-(4-azidophenyl)prop-2-enal
C1=CC(=CC=C1C=CC=O)N=[N+]=[N-]
Color Undesignated
Quantity 5g
Melting Point 75°C
UN Number 1325
Formula Weight 173.18
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (HPLC)
Chemical Name or Material 4-Azidocinnamaldehyde