Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Benzoyl-3-methyl-1-phenyl-5-pyrazolinone, 98+%

Click to view available options
Quantity:
10g
50g
4551-69-3
C17H14N2O2
MFCD00003136
4-Benzoyl-2,4-dihydro-5-methyl-2-phenyl-3H-pyrazol-3-one
4551-69-3
C17H14N2O2
MFCD00003136
4-Benzoyl-2,4-dihydro-5-methyl-2-phenyl-3H-pyrazol-3-one
Melting Point 90°C to 92°C
Quantity 10g
Beilstein 888605
Formula Weight 278.31
Percent Purity ≥98%
Chemical Name or Material 4-Benzoyl-3-methyl-1-phenyl-5-pyrazolinone