Learn More
4-Benzoyl-L-phenylalanine, 95%, Thermo Scientific Chemicals
Description
4-Benzoyl-L-phenylalanine is a photoreactive amino acid which can easily be incorporated into a peptide by solid-phase synthesis and finds application as phtolabel. It is an important raw material as well as an intermediate in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
4-Benzoyl-L-phenylalanine is a photoreactive amino acid which can easily be incorporated into a peptide by solid-phase synthesis and finds application as phtolabel. It is an important raw material as well as an intermediate in organic synthesis.
Notes
Store in a cool place. Incompatible with strong oxidizing agents.
Specifications
Formula Weight | 269.3 |
Percent Purity | 95% |
Quantity | 1 g |
Chemical Name or Material | 4-Benzoyl-L-phenylalanine |
Safety and Handling
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.