Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Benzoyloxy-2-azetidinone 98.0+%, TCI America™
SDP

Catalog No. b13915g Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B13915G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B13915G Supplier TCI America Supplier No. B13915G
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 28562-58-5
Molecular Formula C10H9NO3
Molecular Weight (g/mol) 191.19
MDL Number MFCD00012317
InChI Key HJJGOOONOIFDRH-VIFPVBQESA-N
PubChem CID 2724624
IUPAC Name (2S)-4-oxoazetidin-2-yl benzoate
SMILES O=C(O[C@H]1CC(=O)N1)C1=CC=CC=C1

Specifications

Melting Point 100°C
Color White-Yellow
Quantity 5 g
Formula Weight 191.19
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 4-Benzoyloxy-2-azetidinone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.