Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Benzoylpyridine, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
Packaging:
Glass bottle
Plastic bottle
14548-46-0
C12H9NO
183.21
MFCD00006430
SKFLCXNDKRUHTA-UHFFFAOYSA-N
4-benzoylpyridine,phenyl pyridin-4-yl methanone,phenyl 4-pyridyl ketone,methanone, phenyl-4-pyridinyl,phenyl 4-pyridinyl methanone,4-pyridyl phenyl ketone,pyridine, 4-benzoyl,ketone, phenyl 4-pyridyl,gamma-benzoylpyridine,4-benzoyl pyridine
26731
phenyl(pyridin-4-yl)methanone
C1=CC=C(C=C1)C(=O)C2=CC=NC=C2
14548-46-0
C12H9NO
183.21
MFCD00006430
SKFLCXNDKRUHTA-UHFFFAOYSA-N
4-benzoylpyridine,phenyl pyridin-4-yl methanone,phenyl 4-pyridyl ketone,methanone, phenyl-4-pyridinyl,phenyl 4-pyridinyl methanone,4-pyridyl phenyl ketone,pyridine, 4-benzoyl,ketone, phenyl 4-pyridyl,gamma-benzoylpyridine,4-benzoyl pyridine
26731
phenyl(pyridin-4-yl)methanone
C1=CC=C(C=C1)C(=O)C2=CC=NC=C2
Color Yellow
Boiling Point 315.0°C
Flash Point 150°C
Melting Point 68.0°C to 72.0°C
Infrared Spectrum Authentic
Quantity 100g
Packaging Plastic bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Linear Formula C6H5CO(C5H4N)
Beilstein 21, 331
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in methanol,very soluble in ether & ethanol
Formula Weight 183.21
Physical Form Crystalline Powder or Crystalline Chunks
Percent Purity 98%
Chemical Name or Material 4-Benzoylpyridine, 98%
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS Signal Word: Warning

EINECSNumber : 238-586-7

RTECSNumber : OB6402000

TSCA : TSCA