Alfa Aesar™ 4-Benzylamino-1-butanol, 98+%

Manufacturer: Alfa Aesar™ H30821-18

Quantity 50g

 View more versions of this product

Catalog No. AAH3082118

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 59578-63-1
    Molecular Formula C11H17NO
    Linear Formula C6H5CH2NH(CH2)4OH
    Formula Weight 179.26
    MDL Number MFCD00191609
    Chemical Name or Material 4-Benzylamino-1-butanol

    Specifications

    Formula Weight 179.26
    Molecular Formula C11H17NO
    Linear Formula C6H5CH2NH(CH2)4OH
    MDL Number MFCD00191609
    CAS 59578-63-1
    Boiling Point 140°C (0.8mmHg)
    Health Hazard 1 Warning
    Quantity 50g
    Health Hazard 2 GHS H Statement
    H315-H319
    Causes skin irritation.
    Causes serious eye irritation.
    Health Hazard 3 GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Chemical Name or Material 4-Benzylamino-1-butanol
    Assay ≥98%

    GHS H Statement
    H315-H319
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

    Warning

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit