Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Benzylbenzeneboronic acid pinacol ester, 97%

Manufacturer:  Alfa Aesar™ H2815606

Quantity 5g

 View more versions of this product

Catalog No. AAH2815606


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

911708-01-5
294.2
4-Benzylphenylboronic acid pinacol ester; 2-(4-Benzylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
C19H23BO2
MFCD09266180
4-Benzylbenzeneboronic acid pinacol ester

Specifications

911708-01-5
294.2
4-Benzylphenylboronic acid pinacol ester; 2-(4-Benzylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
5g
97%
C19H23BO2
MFCD09266180
90° to 93°C
4-Benzylbenzeneboronic acid pinacol ester
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit