Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4'-Benzyloxy-2'-hydroxyacetophenone, 98%, Acros Organics

Click to view available options
Quantity:
1g
29682-12-0
C15H14O3
242.28
AGQNLHOTLJFJCG-UHFFFAOYSA-N
1-4-benzyloxy-2-hydroxyphenyl ethanone,4'-benzyloxy-2'-hydroxyacetophenone,4-benzyloxy-2-hydroxyacetophenone,2-hydroxy-4-benzyloxyacetophenone,ethanone, 1-2-hydroxy-4-phenylmethoxy phenyl,1-2-hydroxy-4-phenylmethoxyphenyl ethanone,1-2-hydroxy-4-phenylmethoxy-phenyl ethanone,1-4-benzyloxy-2-hydroxyphenyl ethan-1-one
309257
1-(2-hydroxy-4-phenylmethoxyphenyl)ethanone
CC(=O)C1=C(C=C(C=C1)OCC2=CC=CC=C2)O
29682-12-0
C15H14O3
242.28
AGQNLHOTLJFJCG-UHFFFAOYSA-N
1-4-benzyloxy-2-hydroxyphenyl ethanone,4'-benzyloxy-2'-hydroxyacetophenone,4-benzyloxy-2-hydroxyacetophenone,2-hydroxy-4-benzyloxyacetophenone,ethanone, 1-2-hydroxy-4-phenylmethoxy phenyl,1-2-hydroxy-4-phenylmethoxyphenyl ethanone,1-2-hydroxy-4-phenylmethoxy-phenyl ethanone,1-4-benzyloxy-2-hydroxyphenyl ethan-1-one
309257
1-(2-hydroxy-4-phenylmethoxyphenyl)ethanone
CC(=O)C1=C(C=C(C=C1)OCC2=CC=CC=C2)O
Packaging Glass Bottle
Melting Point 104.0°C to 106.0°C
Infrared Spectrum Authentic
Quantity 1g
Assay Percent Range 97.5% min. (HPLC)
Formula Weight 242.28
Percent Purity 98%
Chemical Name or Material 4'-Benzyloxy-2'-hydroxyacetophenone