Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Benzyloxy-3,5-dimethylbenzaldehyde, 95%

Click to view available options
Quantity:
1g
5g
144896-51-5
C16H16O2
240.302
MFCD00800683
GSYUTKRSEZMBNC-UHFFFAOYSA-N
4-benzyloxy-3,5-dimethylbenzaldehyde,3,5-dimethyl-4-phenylmethoxy benzaldehyde,zlchem 1072,acmc-1bxy6,3,5-dimethyl-4-benzyloxybenzaldehyde,3,5-dimethyl-4-benzyloxy benzaldehyde,4-benzyloxy-3,5-dimethyl-benzaldehyde
563557
3,5-dimethyl-4-phenylmethoxybenzaldehyde
CC1=CC(=CC(=C1OCC2=CC=CC=C2)C)C=O
144896-51-5
C16H16O2
240.302
MFCD00800683
GSYUTKRSEZMBNC-UHFFFAOYSA-N
4-benzyloxy-3,5-dimethylbenzaldehyde,3,5-dimethyl-4-phenylmethoxy benzaldehyde,zlchem 1072,acmc-1bxy6,3,5-dimethyl-4-benzyloxybenzaldehyde,3,5-dimethyl-4-benzyloxy benzaldehyde,4-benzyloxy-3,5-dimethyl-benzaldehyde
563557
3,5-dimethyl-4-phenylmethoxybenzaldehyde
CC1=CC(=CC(=C1OCC2=CC=CC=C2)C)C=O
Quantity 5g
Flash Point >110°C (230°F)
Boiling Point 162°C to 163°C (1mmHg)
Formula Weight 240.3
Percent Purity 95%
Chemical Name or Material 4-Benzyloxy-3,5-dimethylbenzaldehyde