Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Benzyloxy-3,5-dimethylbenzaldehyde, ≥98%, Acros Organics

Click to view available options
Quantity:
5g
144896-51-5
C16H16O2
240.302
MFCD00800683
GSYUTKRSEZMBNC-UHFFFAOYSA-N
4-benzyloxy-3,5-dimethylbenzaldehyde,4-benzyloxy-3,5-dimethylbenzaldehyde,3,5-dimethyl-4-phenylmethoxy benzaldehyde,zlchem 1072,acmc-1bxy6,3,5-dimethyl-4-benzyloxybenzaldehyde,3,5-dimethyl-4-benzyloxy benzaldehyde,4-benzyloxy-3,5-dimethyl-benzaldehyde,3,5-dimethyl-4-benzyloxy benzaldehyde,4-benzyloxy-3,5-dimethyl-benzaldehyde
563557
3,5-dimethyl-4-phenylmethoxybenzaldehyde
CC1=CC(=CC(=C1OCC2=CC=CC=C2)C)C=O
144896-51-5
C16H16O2
240.302
MFCD00800683
GSYUTKRSEZMBNC-UHFFFAOYSA-N
4-benzyloxy-3,5-dimethylbenzaldehyde,4-benzyloxy-3,5-dimethylbenzaldehyde,3,5-dimethyl-4-phenylmethoxy benzaldehyde,zlchem 1072,acmc-1bxy6,3,5-dimethyl-4-benzyloxybenzaldehyde,3,5-dimethyl-4-benzyloxy benzaldehyde,4-benzyloxy-3,5-dimethyl-benzaldehyde,3,5-dimethyl-4-benzyloxy benzaldehyde,4-benzyloxy-3,5-dimethyl-benzaldehyde
563557
3,5-dimethyl-4-phenylmethoxybenzaldehyde
CC1=CC(=CC(=C1OCC2=CC=CC=C2)C)C=O
Quantity 5g
Infrared Spectrum Authentic
Color Yellow
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98+%
Linear Formula C6H5CHOC6H(CH3)CH(O)
Formula Weight 240.3
Physical Form Viscous Liquid After Melting
Percent Purity ≥98.0%
Chemical Name or Material 4-Benzyloxy-3, 5-dimethylbenzaldehyde, 98+%
Show More Show Less