Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Benzyloxy-3-methoxybenzyl alcohol, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
33693-48-0
C15H16O3
244.29
MFCD00004640
PDBXFVPMVYQICB-UHFFFAOYSA-N
4-benzyloxy-3-methoxybenzyl alcohol,4-benzyloxy-3-methoxyphenyl methanol,4-benzyloxy-3-methoxybenzylalcohol,benzenemethanol, 3-methoxy-4-phenylmethoxy,acmc-20amos,o-benzylvanillyl alcohol,4-o-benzyl vanillyl alcohol,vanillyl alcohol benzyl ether
98832
(3-methoxy-4-phenylmethoxyphenyl)methanol
COC1=C(C=CC(=C1)CO)OCC2=CC=CC=C2
33693-48-0
C15H16O3
244.29
MFCD00004640
PDBXFVPMVYQICB-UHFFFAOYSA-N
4-benzyloxy-3-methoxybenzyl alcohol,4-benzyloxy-3-methoxyphenyl methanol,4-benzyloxy-3-methoxybenzylalcohol,benzenemethanol, 3-methoxy-4-phenylmethoxy,acmc-20amos,o-benzylvanillyl alcohol,4-o-benzyl vanillyl alcohol,vanillyl alcohol benzyl ether
98832
(3-methoxy-4-phenylmethoxyphenyl)methanol
COC1=C(C=CC(=C1)CO)OCC2=CC=CC=C2
Infrared Spectrum Authentic
Color White
Melting Point 70°C to 73°C
Packaging Glass bottle
Quantity 1g
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula C6H5CH2OC6H3(OCH3)CH2OH
Formula Weight 244.29
Physical Form Crystalline Powder
Percent Purity ≥96%
Chemical Name or Material 4-Benzyloxy-3-methoxybenzyl alcohol, 97%
Show More Show Less

EINECSNumber : 251-638-3