Alfa Aesar™ 4-Benzyloxy-5-bromo-2-chloropyrimidine, 97%

Manufacturer: Alfa Aesar™ H27706MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH27706MD

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 205672-19-1
    Molecular Formula C11H8BrClN2O
    Formula Weight 299.56
    MDL Number MFCD06798237
    Chemical Name or Material 4-Benzyloxy-5-bromo-2-chloropyrimidine

    Specifications

    Formula Weight 299.56
    Molecular Formula C11H8BrClN2O
    MDL Number MFCD06798237
    CAS 205672-19-1
    Melting Point 114° to 116°C
    Quantity 250mg
    Chemical Name or Material 4-Benzyloxy-5-bromo-2-chloropyrimidine
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit