Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Benzyloxyaniline hydrochloride, 98%

Click to view available options
Quantity:
25g
100g
51388-20-6
C13H14ClNO
235.711
MFCD00012995
KQBDLOVXZHOAJI-UHFFFAOYSA-N
4-benzyloxyaniline hydrochloride,4-benzyloxy aniline hydrochloride,4-benzyloxyaniline hcl,benzenamine, 4-phenylmethoxy-, hydrochloride,4-phenylmethoxyaniline hydrochloride,aniline, p-benzyloxy-hydrochloride,4-benzyloxy phenyl amine hydrochloride,4-phenylmethoxy phenylamine, chloride,acmc-20amop,pubchem21343
2723831
4-phenylmethoxyaniline;hydrochloride
C1=CC=C(C=C1)COC2=CC=C(C=C2)N.Cl
51388-20-6
C13H14ClNO
235.711
MFCD00012995
KQBDLOVXZHOAJI-UHFFFAOYSA-N
4-benzyloxyaniline hydrochloride,4-benzyloxy aniline hydrochloride,4-benzyloxyaniline hcl,benzenamine, 4-phenylmethoxy-, hydrochloride,4-phenylmethoxyaniline hydrochloride,aniline, p-benzyloxy-hydrochloride,4-benzyloxy phenyl amine hydrochloride,4-phenylmethoxy phenylamine, chloride,acmc-20amop,pubchem21343
2723831
4-phenylmethoxyaniline;hydrochloride
C1=CC=C(C=C1)COC2=CC=C(C=C2)N.Cl
Quantity 25g
Melting Point ~225°C (decomposition)
Beilstein 3633307
Formula Weight 235.72
Percent Purity 98%
Chemical Name or Material 4-Benzyloxyaniline hydrochloride

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 257-170-6

RTECSNumber : BW7615000

TSCA : No

Recommended Storage : Ambient temperatures