Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Benzyloxyaniline hydrochloride, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Plastic bottle
51388-20-6
C13H13NO·HCl
235.72
MFCD00012995
KQBDLOVXZHOAJI-UHFFFAOYSA-N
4-benzyloxyaniline hydrochloride,4-benzyloxy aniline hydrochloride,4-benzyloxyaniline hcl,benzenamine, 4-phenylmethoxy-, hydrochloride,4-phenylmethoxyaniline hydrochloride,aniline, p-benzyloxy-hydrochloride,4-benzyloxy phenyl amine hydrochloride,4-phenylmethoxy phenylamine, chloride,acmc-20amop,pubchem21343
2723831
4-phenylmethoxyaniline;hydrochloride
C1=CC=C(C=C1)COC2=CC=C(C=C2)N.Cl
51388-20-6
C13H13NO·HCl
235.72
MFCD00012995
KQBDLOVXZHOAJI-UHFFFAOYSA-N
4-benzyloxyaniline hydrochloride,4-benzyloxy aniline hydrochloride,4-benzyloxyaniline hcl,benzenamine, 4-phenylmethoxy-, hydrochloride,4-phenylmethoxyaniline hydrochloride,aniline, p-benzyloxy-hydrochloride,4-benzyloxy phenyl amine hydrochloride,4-phenylmethoxy phenylamine, chloride,acmc-20amop,pubchem21343
2723831
4-phenylmethoxyaniline;hydrochloride
C1=CC=C(C=C1)COC2=CC=C(C=C2)N.Cl
Melting Point 228°C
Quantity 25g
Packaging Plastic bottle
Infrared Spectrum Authentic
Color Brown
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula C6H5CH2OC6H4NH2·HCl
Beilstein 13, 439
Solubility Information Solubility in water: slightly soluble
Formula Weight 235.72
Physical Form Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material 4-Benzyloxyaniline hydrochloride, 98%
Show More Show Less

GHS P Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 257-170-6