Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Benzyloxybenzaldehyde, 98%, Thermo Scientific Chemicals

Catalog No. p-65536
Click to view available options
Quantity:
100 g
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4397-53-9
Molecular Formula C14H12O2
Molecular Weight (g/mol) 212.25
MDL Number MFCD00003387,MFCD01632880
InChI Key ZVTWZSXLLMNMQC-UHFFFAOYSA-N
Synonym 4-benzyloxybenzaldehyde, 4-benzyloxy benzaldehyde, benzaldehyde, 4-phenylmethoxy, p-benzyloxy benzaldehyde, benzaldehyde, p-benzyloxy, 4-benzyloxy-benzaldehyde, 4-phenylmethoxy benzaldehyde, 4-benzyl-oxy benzaldehyde, wang aldehyde resin
PubChem CID 78109
IUPAC Name 4-phenylmethoxybenzaldehyde
SMILES O=CC1=CC=C(OCC2=CC=CC=C2)C=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 70°C to 74°C
Color Cream to Yellow
Boiling Point 197°C to 199°C (11.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5CH2OC6H4CHO
Quantity 100 g
Beilstein 08, 73
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol and chloroform
Formula Weight 212.25
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 4-Benzyloxybenzaldehyde, 98%
Show More Show Less
EINECSNumber 224-527-2
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.