Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Benzyloxybenzeneboronic acid, 97%

Click to view available options
Quantity:
0.25g
1g
5g
146631-00-7
C13H13BO3
228.054
MFCD01075705
DMJHEIDWSIAXCS-UHFFFAOYSA-N
4-benzyloxyphenylboronic acid,4-benzyloxybenzeneboronic acid,4-benzyloxy phenyl boronic acid,4-benzyloxy phenylboronic acid,4-benzyloxyphenyl boronic acid,4-benzyl-oxy phenyl boranediol,boronic acid, 4-phenylmethoxy phenyl,4-phenylmethoxyphenyl boronic acid,pubchem9561
2734314
(4-phenylmethoxyphenyl)boronic acid
B(C1=CC=C(C=C1)OCC2=CC=CC=C2)(O)O
146631-00-7
C13H13BO3
228.054
MFCD01075705
DMJHEIDWSIAXCS-UHFFFAOYSA-N
4-benzyloxyphenylboronic acid,4-benzyloxybenzeneboronic acid,4-benzyloxy phenyl boronic acid,4-benzyloxy phenylboronic acid,4-benzyloxyphenyl boronic acid,4-benzyl-oxy phenyl boranediol,boronic acid, 4-phenylmethoxy phenyl,4-phenylmethoxyphenyl boronic acid,pubchem9561
2734314
(4-phenylmethoxyphenyl)boronic acid
B(C1=CC=C(C=C1)OCC2=CC=CC=C2)(O)O
Applications
It is used as electronic intermediate. It is also utilized in HPLC and NMR spectroscopy.

Notes
Store at room temperature. Incompatible with oxidizing agents.
Quantity 1g
Melting Point 193°C to 198°C
Formula Weight 228.06
Percent Purity 97%
Chemical Name or Material 4-Benzyloxybenzeneboronic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning