Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Benzyloxybenzyl alcohol, 99%, ACROS Organics™

Click to view available options
Quantity:
10g
25g
100g
Packaging:
Glass bottle
836-43-1
C14H14O2
214.26
MFCD00004654
OEBIVOHKFYSBPE-UHFFFAOYSA-N
4-benzyloxybenzyl alcohol,4-benzyloxy phenyl methanol,p-benzyloxybenzyl alcohol,4-benzyloxybenzylalcohol,4-benzyloxy benzyl alcohol,benzenemethanol, 4-phenylmethoxy,p-alkoxybenzyl alcohol resin,4-phenylmethoxyphenyl methanol,argopore-wang
70043
CHEBI:29486
(4-phenylmethoxyphenyl)methanol
C1=CC=C(C=C1)COC2=CC=C(C=C2)CO
836-43-1
C14H14O2
214.26
MFCD00004654
OEBIVOHKFYSBPE-UHFFFAOYSA-N
4-benzyloxybenzyl alcohol,4-benzyloxy phenyl methanol,p-benzyloxybenzyl alcohol,4-benzyloxybenzylalcohol,4-benzyloxy benzyl alcohol,benzenemethanol, 4-phenylmethoxy,p-alkoxybenzyl alcohol resin,4-phenylmethoxyphenyl methanol,argopore-wang
70043
CHEBI:29486
(4-phenylmethoxyphenyl)methanol
C1=CC=C(C=C1)COC2=CC=C(C=C2)CO
Quantity 25g
Color White
Packaging Glass bottle
Infrared Spectrum Authentic
Melting Point 84°C to 87°C
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula C6H5CH2OC6H4CH2OH
Formula Weight 214.26
Physical Form Powder
Percent Purity 99%
Chemical Name or Material 4-Benzyloxybenzyl alcohol
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 212-649-9