Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Benzyloxybromobenzene, 97%, Acros Organics

Click to view available options
Quantity:
10g
50g
6793-92-6
C13H11BrO
263.14
OUQSGILAXUXMGI-UHFFFAOYSA-N
4-benzyloxybromobenzene,1-benzyloxy-4-bromobenzene,1-bromo-4-phenylmethoxy benzene,1-bromo-4-benzyloxybenzene,4-benzyloxy bromobenzene,1-benzyloxy-4-bromo-benzene,1-bromo-4-phenylmethyl oxy benzene,4-benzyloxy-bromobenzene,p-benzyloxy bromobenzene
138835
1-bromo-4-phenylmethoxybenzene
C1=CC=C(C=C1)COC2=CC=C(C=C2)Br
6793-92-6
C13H11BrO
263.14
OUQSGILAXUXMGI-UHFFFAOYSA-N
4-benzyloxybromobenzene,1-benzyloxy-4-bromobenzene,1-bromo-4-phenylmethoxy benzene,1-bromo-4-benzyloxybenzene,4-benzyloxy bromobenzene,1-benzyloxy-4-bromo-benzene,1-bromo-4-phenylmethyl oxy benzene,4-benzyloxy-bromobenzene,p-benzyloxy bromobenzene
138835
1-bromo-4-phenylmethoxybenzene
C1=CC=C(C=C1)COC2=CC=C(C=C2)Br
Boiling Point 166°C to 167°C (4.0 Torr)
Melting Point 62°C to 64°C
Quantity 10g
Assay Percent Range 96% min. (GC)
Formula Weight 263.14
Percent Purity 97%
Chemical Name or Material 4-Benzyloxybromobenzene

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and

GHS Signal Word: Warning