Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-(Benzyloxycarbonylamino)butyric acid, 98%

Click to view available options
Quantity:
5g
25g
100g
5105-78-2
C12H15NO4
237.255
MFCD00055835
STQMDRQJSNKUAW-UHFFFAOYSA-N
z-gamma-abu-oh,4-benzyloxy carbonyl amino butanoic acid,n-cbz-gaba,4-benzyloxy carbonyl amino butanoic acid,carbobenzoxy-4-aminobutyric acid,n-cbz-4-aminobutanoic acid,z-gaba-oh,n-cbz-4-aminobutyric acid,4-carboxyamino butyric acid n-benzyl ester,z-l-gamma-aminobutyric acid
21184
4-(phenylmethoxycarbonylamino)butanoic acid
C1=CC=C(C=C1)COC(=O)NCCCC(=O)O
5105-78-2
C12H15NO4
237.255
MFCD00055835
STQMDRQJSNKUAW-UHFFFAOYSA-N
z-gamma-abu-oh,4-benzyloxy carbonyl amino butanoic acid,n-cbz-gaba,4-benzyloxy carbonyl amino butanoic acid,carbobenzoxy-4-aminobutyric acid,n-cbz-4-aminobutanoic acid,z-gaba-oh,n-cbz-4-aminobutyric acid,4-carboxyamino butyric acid n-benzyl ester,z-l-gamma-aminobutyric acid
21184
4-(phenylmethoxycarbonylamino)butanoic acid
C1=CC=C(C=C1)COC(=O)NCCCC(=O)O
Quantity 25g
Beilstein 1884426
Formula Weight 237.26
Physical Form Solid
Percent Purity 98%
Chemical Name or Material 4-(Benzyloxycarbonylamino)butyric acid

RTECSNumber : ES8137200