Alfa Aesar™ 4-(Benzyloxycarbonylamino)cyclohexanol, 97%, predominantly trans

Manufacturer: Alfa Aesar™ H2601614

Quantity 25g

 View more versions of this product

Catalog No. AAH2601614

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 16801-62-0
    Molecular Formula C14H19NO3
    Formula Weight 249.31
    MDL Number MFCD02728352
    Synonym 4-(Cbz-amino)cyclohexanol; N-Benzyloxycarbonyl-4-hydroxycyclohexylamine
    Chemical Name or Material 4-(Benzyloxycarbonylamino)cyclohexanol, predominantly trans

    Specifications

    Formula Weight 249.31
    Synonym 4-(Cbz-amino)cyclohexanol; N-Benzyloxycarbonyl-4-hydroxycyclohexylamine
    Quantity 25g
    Molecular Formula C14H19NO3
    MDL Number MFCD02728352
    CAS 16801-62-0
    Melting Point 162° to 164°C
    Odor Odorless
    Health Hazard 2 GHS H Statement
    H302
    Harmful if swallowed.
    Health Hazard 3 GHS P Statement
    P280f
    Wear protective clothing.
    Health Hazard 1 Warning
    Chemical Name or Material 4-(Benzyloxycarbonylamino)cyclohexanol, predominantly trans
    Assay 97%

    GHS H Statement
    H302
    Harmful if swallowed.

    GHS P Statement
    P280f
    Wear protective clothing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit