Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Benzyloxyindole, 98%, Acros Organics

Click to view available options
Quantity:
1g
20289-26-3
C15H13NO
223.275
MFCD00047200
LJFVSIDBFJPKLD-UHFFFAOYSA-N
4-benzyloxyindole,4-benzyloxy-1h-indole,4-phenylmethoxy-1h-indole,1h-indole, 4-phenylmethoxy,4-benzyloxy-indole,4-phenylmethoxy indole,4-benzoxyindole,4-benzyloxy-indol,4-benzyloxyindole;
88465
4-phenylmethoxy-1H-indole
C1=CC=C(C=C1)COC2=CC=CC3=C2C=CN3
20289-26-3
C15H13NO
223.275
MFCD00047200
LJFVSIDBFJPKLD-UHFFFAOYSA-N
4-benzyloxyindole,4-benzyloxy-1h-indole,4-phenylmethoxy-1h-indole,1h-indole, 4-phenylmethoxy,4-benzyloxy-indole,4-phenylmethoxy indole,4-benzoxyindole,4-benzyloxy-indol,4-benzyloxyindole;
88465
4-phenylmethoxy-1H-indole
C1=CC=C(C=C1)COC2=CC=CC3=C2C=CN3
Color White
Quantity 1g
Infrared Spectrum Authentic
Melting Point 57°C to 61°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 223.38
Physical Form Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material 4-Benzyloxyindole
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.