Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Benzylpiperazine-1-carboximidamide hydroiodide, 95%, Thermo Scientific™

Click to view available options
Quantity:
1g
10g
Packaging:
Amber Glass Bottle
852228-15-0
C12H19IN4
346.22
MFCD09025921
CXQITJKGJPFOIV-UHFFFAOYSA-N
4-benzylpiperazine-1-carboximidamide hydroiodide, 4-benzylpiperazine-1-carboximidamide-hydrogen iodide 1/1
18525929
4-benzylpiperazine-1-carboximidamide;hydroiodide
I.NC(=N)N1CCN(CC2=CC=CC=C2)CC1
852228-15-0
C12H19IN4
346.22
MFCD09025921
CXQITJKGJPFOIV-UHFFFAOYSA-N
4-benzylpiperazine-1-carboximidamide hydroiodide, 4-benzylpiperazine-1-carboximidamide-hydrogen iodide 1/1
18525929
4-benzylpiperazine-1-carboximidamide;hydroiodide
I.NC(=N)N1CCN(CC2=CC=CC=C2)CC1
CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 10g
Formula Weight 346.21
Percent Purity 95%
Chemical Name or Material 4-Benzylpiperazine-1-carboximidamide hydroiodide

GHS H Statement:
Causes skin irritation.
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning