Alfa Aesar™ 4-Benzylpiperidine, 98%

Manufacturer: Alfa Aesar™ B2388314

Quantity 25g

 View more versions of this product

Catalog No. AAB2388314

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 31252-42-3
    Molecular Formula C12H17N
    Formula Weight 175.28
    MDL Number MFCD00006006
    Beilstein 132339
    Chemical Name or Material 4-Benzylpiperidine

    Specifications

    Formula Weight 175.28
    TSCA TSCA
    Molecular Formula C12H17N
    Melting Point 6° to 7°C
    MDL Number MFCD00006006
    Beilstein 132339
    Boiling Point 278° to 279°C
    Quantity 25g
    Flash Point 147°C (296°F)
    Health Hazard 2 GHS H Statement
    H315-H319
    Causes skin irritation.
    Causes serious eye irritation.
    Health Hazard 3 GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    RTECS Number TM4728000
    CAS 31252-42-3
    Health Hazard 1 Warning
    Chemical Name or Material 4-Benzylpiperidine
    Assay 98%

    GHS H Statement
    H315-H319
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit