Alfa Aesar™ 4-Biphenylacetic acid, 98%

Manufacturer: Alfa Aesar™ A1174930

Quantity 250g

 View more versions of this product

Catalog No. AAA1174930

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 5728-52-9
    Molecular Formula C14H12O2
    Formula Weight 212.25
    MDL Number MFCD00004351
    UN Number UN2811
    Beilstein 1211592
    Merck Index 14,3951
    Synonym Felbinac
    Chemical Name or Material 4-Biphenylacetic acid

    Specifications

    Formula Weight 212.25
    Health Hazard 3 GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Merck Index 14,3951
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
    RTECS Number DU8229050
    Molecular Formula C14H12O2
    MDL Number MFCD00004351
    CAS 5728-52-9
    UN Number UN2811
    Beilstein 1211592
    Health Hazard 2 GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Odor Odorless
    Melting Point 158° to 163°C
    Synonym Felbinac
    Quantity 250g
    Health Hazard 1 Danger
    Chemical Name or Material 4-Biphenylacetic acid
    Assay 98%

    GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit