Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Biphenylacetic acid, 98%, ACROS Organics™

Click to view available options
Quantity:
10g
25g
100g
500g
Packaging:
Glass bottle
5728-52-9
C14H12O2
212.25
MFCD00004351
QRZAKQDHEVVFRX-UHFFFAOYSA-N
4-biphenylacetic acid,felbinac,4-biphenylylacetic acid,traxam,dolinac,4-carboxymethylbiphenyl,napageln,2-4-phenylphenyl acetic acid,p-biphenylylacetic acid,1,1'-biphenyl-4-acetic acid
3332
CHEBI:31597
2-(4-phenylphenyl)acetic acid
C1=CC=C(C=C1)C2=CC=C(C=C2)CC(=O)O
5728-52-9
C14H12O2
212.25
MFCD00004351
QRZAKQDHEVVFRX-UHFFFAOYSA-N
4-biphenylacetic acid,felbinac,4-biphenylylacetic acid,traxam,dolinac,4-carboxymethylbiphenyl,napageln,2-4-phenylphenyl acetic acid,p-biphenylylacetic acid,1,1'-biphenyl-4-acetic acid
3332
CHEBI:31597
2-(4-phenylphenyl)acetic acid
C1=CC=C(C=C1)C2=CC=C(C=C2)CC(=O)O
Quantity 100g
Packaging Glass bottle
Infrared Spectrum Authentic
Color Beige
Melting Point 156°C to 160°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula C6H5C6H4CH2CO2H
Beilstein 09, I, 284
Merck Index 15, 3985
Formula Weight 212.25
Physical Form Powder
Percent Purity 98%
Chemical Name or Material 4-Biphenylacetic acid
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 227-233-2